Isopropyl 2-chloronicotinate ≥95%

Supplier: AOBChem USA

Warning

290052-5G 290052-1G 290052-25G 290052-500MG
77632-490EA 1349 USD
77632-490 77637-822 77654-752 77645-778
Isopropyl 2-chloronicotinate ≥95%
Isopropyl 2-chloronicotinate

Precaución: For Research Use Only

Formula: 199.634 g/mol
MW: C₉H₁₀ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD18839830
CAS Number: 152523-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR