4-(3-Bromophenyl)-3,6-dihydro-2H-pyran ≥95%

Supplier: AOBChem USA

Warning

29005-1G 29005-5G 29005-500MG
77634-636EA 1078 USD
77634-636 77654-758 77649-848
4-(3-Bromophenyl)-3,6-dihydro-2H-pyran ≥95%
4-(3-Bromophenyl)-3,6-dihydro-2H-pyran

Precaución: For Research Use Only

Formula: 239.108 g/mol
MW: C₁₁H₁₁BrO
Storage Temperature: Ambient
CAS Number: 1174324-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR