6-Bromo-2-naphthonitrile ≥95%

Supplier: AOBChem USA

Warning

290029-5G 290029-25G 290029-1G 290029-500MG
77634-634EA 593 USD
77634-634 77658-492 77641-774 77645-776
6-Bromo-2-naphthonitrile ≥95%
6-Bromo-2-naphthonitrile

Precaución: For Research Use Only

Formula: C₁₁H₆BrN
Storage Temperature: Ambient
MDL Number: MFCD16620251
CAS Number: 91065-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR