2-(3,6-Difluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

28930-5G 28930-1G 28930-25G 28930-500MG
77634-630EA 594 USD
77634-630 77637-820 77654-748 77645-770
2-(3,6-Difluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3,6-Difluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: For Research Use Only

Formula: C₁₃H₁₇BF₂O₂
MW: 254.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669571
CAS Number: 1025707-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR