4,5-Dichloro-2-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

28866-5G 28866-25G 28866-1G 28866-500MG
77632-484EA 1430 USD
77632-484 77654-746 77641-770 77649-844
4,5-Dichloro-2-(trifluoromethyl)phenylboronic acid ≥95%
4,5-Dichloro-2-(trifluoromethyl)phenylboronic acid

Precaución: For Research Use Only

Formula: 258.818 g/mol
MW: C₇H₄BCl₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD28388514
CAS Number: 2121511-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR