1,4-Dibromo-2-(difluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

288169-5G 288169-1G 288169-500MG
77628-438EA 1484.23 USD
77628-438 77641-768 77649-842
1,4-Dibromo-2-(difluoromethyl)benzene ≥95%
1,4-Dibromo-2-(difluoromethyl)benzene

Precaución: For Research Use Only

Formula: C₇H₄Br₂F₂
Storage Temperature: Ambient
MDL Number: MFCD14698561
CAS Number: 1214383-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR