3,5-Dimethoxy-N,N-dimethylbenzenamine ≥97%

Supplier: AOBChem USA

Warning

287797-5G 287797-1G 287797-500MG
77654-744EA 2417 USD
77654-744 77645-764 77645-766
3,5-Dimethoxy-N,N-dimethylbenzenamine ≥97%
3,5-Dimethoxy-N,N-dimethylbenzenamine

Precaución: For Research Use Only

Formula: C₁₀H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD22055927
CAS Number: 2570-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR