2-Chloro-1-ethyl-3-methyl-5-(methylthio)benzene ≥95%

Supplier: AOBChem USA

Warning

287386-5G 287386-1G 287386-25G 287386-500MG
77628-436EA 1583.11 USD
77628-436 77637-812 77654-738 77649-838
2-Chloro-1-ethyl-3-methyl-5-(methylthio)benzene ≥95%
2-Chloro-1-ethyl-3-methyl-5-(methylthio)benzene

Precaución: For Research Use Only

Formula: 200.73 g/mol
MW: C₁₀H₁₃ClS
Storage Temperature: Ambient
MDL Number: MFCD34886050
CAS Number: 2708456-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR