(4-Cyclopentylphenyl)cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

287216-5G 287216-1G 287216-500MG
77654-736EA 2245 USD
77654-736 77641-764 77649-834
(4-Cyclopentylphenyl)cyclopropylmethanone ≥95%
(4-Cyclopentylphenyl)cyclopropylmethanone

Precaución: For Research Use Only

Formula: 214.3 g/mol
MW: C₁₅H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD34885968
CAS Number: 1506525-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR