Cyclopropyl(3-methoxy-4-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287183-5G 287183-1G 287183-500MG
77628-434EA 1599.59 USD
77628-434 77641-762 77649-832
Cyclopropyl(3-methoxy-4-methylphenyl)methanone ≥95%
Cyclopropyl(3-methoxy-4-methylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD29942271
CAS Number: 1592327-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR