(3-Chloro-2,6-difluorophenyl)cyclopropylmethanone ≥97%

Supplier: AOBChem USA

Warning

287164-1G 287164-5G 287164-500MG
77637-808EA 591 USD
77637-808 77654-734 77649-830
(3-Chloro-2,6-difluorophenyl)cyclopropylmethanone ≥97%
(3-Chloro-2,6-difluorophenyl)cyclopropylmethanone

Precaución: For Research Use Only

Formula: C₁₀H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD34885938
CAS Number: 2353115-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR