Cyclopropyl(2,6-difluoro-3-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287150-1G 287150-5G 287150-500MG
77637-806EA 531 USD
77637-806 77654-732 77649-828
Cyclopropyl(2,6-difluoro-3-methylphenyl)methanone ≥95%
Cyclopropyl(2,6-difluoro-3-methylphenyl)methanone

Precaución: For Research Use Only

Formula: 196.19 g/mol
MW: C₁₁H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD24310427
CAS Number: 1551568-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR