(3-Chloro-5-methylphenyl)cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

287146-1G 287146-5G 287146-500MG
77637-804EA 555 USD
77637-804 77654-730 77645-760
(3-Chloro-5-methylphenyl)cyclopropylmethanone ≥95%
(3-Chloro-5-methylphenyl)cyclopropylmethanone

Precaución: For Research Use Only

Formula: 194.66 g/mol
MW: C₁₁H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD30163454
CAS Number: 1602724-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR