(2-Chloro-3,6-difluorophenyl)cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

287135-1G 287135-5G 287135-500MG
77637-800EA 507 USD
77637-800 77654-726 77649-826
(2-Chloro-3,6-difluorophenyl)cyclopropylmethanone ≥95%
(2-Chloro-3,6-difluorophenyl)cyclopropylmethanone

Precaución: For Research Use Only

Formula: 216.61 g/mol
MW: C₁₀H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD30628750
CAS Number: 2148478-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR