Cyclopropyl(5-methoxy-2-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287131-5G 287131-1G 287131-500MG
77628-432EA 1599.59 USD
77628-432 77637-798 77645-756
Cyclopropyl(5-methoxy-2-methylphenyl)methanone ≥95%
Cyclopropyl(5-methoxy-2-methylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34885923
CAS Number: 2299998-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR