Cyclopropyl[2-methyl-3-(trifluoromethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

287126-5G 287126-1G 287126-500MG
77658-484EA 1983 USD
77658-484 77641-760 77645-754
Cyclopropyl[2-methyl-3-(trifluoromethyl)phenyl]methanone ≥95%
Cyclopropyl[2-methyl-3-(trifluoromethyl)phenyl]methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₁F₃O
Storage Temperature: Ambient
MDL Number: MFCD28410889
CAS Number: 2090426-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR