(3-Chloro-4-methoxyphenyl)cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

287123-5G 287123-1G 287123-25G 287123-500MG
77632-480EA 1441 USD
77632-480 77637-796 77654-724 77649-824
(3-Chloro-4-methoxyphenyl)cyclopropylmethanone ≥95%
(3-Chloro-4-methoxyphenyl)cyclopropylmethanone

Precaución: For Research Use Only

Formula: 210.66 g/mol
MW: C₁₁H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD02947810
CAS Number: 494780-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR