Cyclopropyl[3-(1-methylethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

287111-1G 287111-5G 287111-500MG
77637-790EA 560 USD
77637-790 77654-722 77645-748
Cyclopropyl[3-(1-methylethyl)phenyl]methanone ≥95%
Cyclopropyl[3-(1-methylethyl)phenyl]methanone

Precaución: For Research Use Only

Formula: C₁₃H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD28408802
CAS Number: 1935110-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR