[3-Chloro-4-(trifluoromethyl)phenyl]cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

287104-1G 287104-5G 287104-500MG
77637-788EA 560 USD
77637-788 77654-718 77649-822
[3-Chloro-4-(trifluoromethyl)phenyl]cyclopropylmethanone ≥95%
[3-Chloro-4-(trifluoromethyl)phenyl]cyclopropylmethanone

Precaución: For Research Use Only

Formula: 248.63 g/mol
MW: C₁₁H₈ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD28411080
CAS Number: 2090430-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR