[1,1'-Biphenyl]-3-ylcyclopropylmethanone ≥97%

Supplier: AOBChem USA

Warning

287097-5G 287097-1G 287097-500MG
77628-428EA 1484.23 USD
77628-428 77637-786 77649-820
[1,1'-Biphenyl]-3-ylcyclopropylmethanone ≥97%
[1,1'-Biphenyl]-3-ylcyclopropylmethanone

Precaución: For Research Use Only

Formula: C₁₆H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD32898611
CAS Number: 185626-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR