Cyclopropyl(3-fluoro-2-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287095-5G 287095-1G 287095-500MG
77658-480EA 1983 USD
77658-480 77641-756 77649-818
Cyclopropyl(3-fluoro-2-methylphenyl)methanone ≥95%
Cyclopropyl(3-fluoro-2-methylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₁H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD28408357
CAS Number: 1422677-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR