Cyclopropyl(2,4,6-trifluorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287094-1G 287094-500MG 287094-5G
77637-784EA 531 USD
77637-784 77652-600 77654-716
Cyclopropyl(2,4,6-trifluorophenyl)methanone ≥95%
Cyclopropyl(2,4,6-trifluorophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₀H₇F₃O
Storage Temperature: Ambient
MDL Number: MFCD11210712
CAS Number: 1094286-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR