(2-Chloro-4-methoxyphenyl)cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

287092-5G 287092-1G 287092-25G 287092-500MG
77632-478EA 1381 USD
77632-478 77637-782 77654-714 77649-816
(2-Chloro-4-methoxyphenyl)cyclopropylmethanone ≥95%
(2-Chloro-4-methoxyphenyl)cyclopropylmethanone

Precaución: For Research Use Only

Formula: 210.66 g/mol
MW: C₁₁H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD12139102
CAS Number: 1156653-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR