Cyclopropyl(4-phenoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287071-5G 287071-1G 287071-25G 287071-500MG
77632-474EA 1381 USD
77632-474 77637-780 77658-474 77645-742
Cyclopropyl(4-phenoxyphenyl)methanone ≥95%
Cyclopropyl(4-phenoxyphenyl)methanone

Precaución: For Research Use Only

Formula: 238.28 g/mol
MW: C₁₆H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD11544942
CAS Number: 1096909-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR