[1,1'-Biphenyl]-2-ylcyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

287066-5G 287066-1G 287066-500MG
77654-712EA 2001 USD
77654-712 77641-748 77645-740
[1,1'-Biphenyl]-2-ylcyclopropylmethanone ≥95%
[1,1'-Biphenyl]-2-ylcyclopropylmethanone

Precaución: For Research Use Only

Formula: C₁₆H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD28406479
CAS Number: 93301-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR