Cyclopropyl(4-methoxy-2-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287064-1G 287064-5G 287064-500MG
77637-778EA 560 USD
77637-778 77654-710 77645-738
Cyclopropyl(4-methoxy-2-methylphenyl)methanone ≥95%
Cyclopropyl(4-methoxy-2-methylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₂H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD21187717
CAS Number: 1484542-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR