Cyclopropyl[3-(1,1-dimethylethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

287062-5G 287062-1G 287062-500MG
77658-472EA 2001 USD
77658-472 77641-746 77649-810
Cyclopropyl[3-(1,1-dimethylethyl)phenyl]methanone ≥95%
Cyclopropyl[3-(1,1-dimethylethyl)phenyl]methanone

Precaución: For Research Use Only

Formula: C₁₄H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD28406636
CAS Number: 1995115-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR