(2-Chloro-4-methylphenyl)cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

287059-5G 287059-1G 287059-500MG
77628-424EA 1484.23 USD
77628-424 77641-744 77649-808
(2-Chloro-4-methylphenyl)cyclopropylmethanone ≥95%
(2-Chloro-4-methylphenyl)cyclopropylmethanone

Precaución: For Research Use Only

Formula: 194.66 g/mol
MW: C₁₁H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD27927128
CAS Number: 66353-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR