Cyclopropyl[3-(phenylmethoxy)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

287058-1G 287058-500MG 287058-5G
77637-776EA 560 USD
77637-776 77652-598 77654-708
Cyclopropyl[3-(phenylmethoxy)phenyl]methanone ≥95%
Cyclopropyl[3-(phenylmethoxy)phenyl]methanone

Precaución: For Research Use Only

Formula: 252.31 g/mol
MW: C₁₇H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD28406386
CAS Number: 1869772-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR