Cyclopropyl(5-fluoro-2-methylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287055-5G 287055-1G 287055-500MG
77654-706EA 2001 USD
77654-706 77645-734 77645-736
Cyclopropyl(5-fluoro-2-methylphenyl)methanone ≥95%
Cyclopropyl(5-fluoro-2-methylphenyl)methanone

Precaución: For Research Use Only

Formula: 178.2 g/mol
MW: C₁₁H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD17222561
CAS Number: 1342070-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR