Cyclopropyl(2,5-dichlorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

287025-5G 287025-25G 287025-1G 287025-500MG
77632-468EA 1268 USD
77632-468 77654-702 77641-742 77649-806
Cyclopropyl(2,5-dichlorophenyl)methanone ≥95%
Cyclopropyl(2,5-dichlorophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₀H₈Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD03841238
CAS Number: 898790-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR