Cyclopropyl-3-trifluoromethylphenyl ketone ≥95%

Supplier: AOBChem USA

Warning

287020-5G 287020-1G 287020-25G
77634-624EA 217 USD
77634-624 77637-768 77658-466
Cyclopropyl-3-trifluoromethylphenyl ketone ≥95%
Cyclopropyl-3-trifluoromethylphenyl ketone

Precaución: For Research Use Only

Formula: C₁₁H₉F₃O
MW: 214.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03841230
CAS Number: 38675-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR