Ethyl 4-(((trifluoromethyl)sulfonyl)oxy)cyclohex-3-enecarboxylate ≥97%
Supplier: AOBChem USA
|
Warning
|
286855-5G
286855-25G
286855-1G
286855-500MG
77632-466EA
1140
USD
77632-466
77654-698
77641-740
77649-804
Ethyl 4-(((trifluoromethyl)sulfonyl)oxy)cyclohex-3-enecarboxylate ≥97%
Ethyl 4-(((trifluoromethyl)sulfonyl)oxy)cyclohex-3-enecarboxylate
Precaución: For Research Use Only
Formula:
302.2674 g/mol MW: C₁₀H₁₃F₃O₅S Storage Temperature: Ambient |
MDL Number:
MFCD17169871 CAS Number: 122948-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...