(2-(Trifluoromethyl)-1,4-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

286832-5G 286832-1G 286832-25G 286832-500MG
77632-464EA 1441 USD
77632-464 77637-766 77658-464 77645-726
(2-(Trifluoromethyl)-1,4-phenylene)bis(methylsulfane) ≥97%
(2-(Trifluoromethyl)-1,4-phenylene)bis(methylsulfane)

Precaución: For Research Use Only

Formula: C₉H₉F₃S₂
MW: 238.2929 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34885821
CAS Number: 2969444-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR