Methyl 4-cyclobutylbenzoate ≥97%

Supplier: AOBChem USA

Warning

28585-5G 28585-1G 28585-500MG
77628-418EA 1484.23 USD
77628-418 77645-724 77649-794
Methyl 4-cyclobutylbenzoate ≥97%
Methyl 4-cyclobutylbenzoate

Precaución: For Research Use Only

Formula: C₁₂H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD19440001
CAS Number: 1378655-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR