Potassium 4-hydroxy-3-methoxyphenyltrifluoroborate ≥97%

Supplier: AOBChem USA

Warning

28528-5G 28528-500MG 28528-25G 28528-1G
77634-616EA 560 USD
77634-616 77652-592 77658-456 77641-728
Potassium 4-hydroxy-3-methoxyphenyltrifluoroborate ≥97%
Potassium 4-hydroxy-3-methoxyphenyltrifluoroborate

Precaución: For Research Use Only

Formula: 230.034 g/mol
MW: C₇H₇BF₃KO₂
Storage Temperature: Ambient
MDL Number: MFCD28360696
CAS Number: 1883442-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR