1-(2-Bromo-4-chlorophenyl)-1H-imidazole ≥97%
Supplier: AOBChem USA
|
Warning
|
28519-5G
28519-1G
28519-25G
28519-500MG
77632-460EA
841
USD
77632-460
77637-764
77654-694
77645-722
1-(2-Bromo-4-chlorophenyl)-1H-imidazole ≥97%
1-(2-Bromo-4-chlorophenyl)-1H-imidazole
Precaución: For Research Use Only
Formula:
C₉H₆BrClN₂ MW: 257.514 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28360694 CAS Number: 1785222-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...