2,3-Dichloro-α-ethenyl-6-fluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

284735-5G 284735-25G 284735-1G 284735-500MG
77628-416EA 1484.23 USD
77628-416 77658-454 77641-726 77645-720
2,3-Dichloro-α-ethenyl-6-fluorobenzenemethanol ≥95%
2,3-Dichloro-α-ethenyl-6-fluorobenzenemethanol

Precaución: For Research Use Only

Formula: 221.06 g/mol
MW: C₉H₇Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD34885157
CAS Number: 149947-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR