2-Methoxy-1-methyl-3-(trimethylsilyl)benzene ≥95%

Supplier: AOBChem USA

Warning

284660-5G 284660-1G 284660-500MG
77628-414EA 1484.23 USD
77628-414 77637-762 77645-718
2-Methoxy-1-methyl-3-(trimethylsilyl)benzene ≥95%
2-Methoxy-1-methyl-3-(trimethylsilyl)benzene

Precaución: For Research Use Only

Formula: 194.35 g/mol
MW: C₁₁H₁₈OSi
Storage Temperature: Ambient
MDL Number: MFCD34885117
CAS Number: 850754-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR