3,5-Diiodopyridine ≥95%

Supplier: AOBChem USA

Warning

28445-5G 28445-1G 28445-25G
77634-614EA 214 USD
77634-614 77637-760 77658-452
3,5-Diiodopyridine ≥95%
3,5-Diiodopyridine

Precaución: For Research Use Only

Formula: C₅H₃I₂N
MW: 330.893 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01928488
CAS Number: 53710-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR