2-(3-Chloro-4-phenoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
Supplier: AOBChem USA
|
Warning
|
28435-5G
28435-25G
28435-1G
28435-500MG
77632-458EA
876
USD
77632-458
77654-692
77641-724
77645-716
2-(3-Chloro-4-phenoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(3-Chloro-4-phenoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: For Research Use Only
Formula:
330.614 g/mol MW: C₁₈H₂₀BClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD22493984 CAS Number: 364354-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...