4-Bromo-2-chloro-1-phenoxybenzene ≥97%
Supplier: AOBChem USA
|
Warning
|
28434-5G
28434-25G
28434-1G
28434-500MG
77632-456EA
915
USD
77632-456
77654-690
77645-712
77645-714
4-Bromo-2-chloro-1-phenoxybenzene ≥97%
4-Bromo-2-chloro-1-phenoxybenzene
Precaución: For Research Use Only
Formula:
283.548 g/mol MW: C₁₂H₈BrClO Storage Temperature: Ambient |
MDL Number:
MFCD28360691 CAS Number: 364354-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...