3,5-Dimethoxy-N-methylaniline ≥97%

Supplier: AOBChem USA

Warning

284120-5G 284120-500MG 284120-25G 284120-1G
77632-454EA 1115 USD
77632-454 77652-590 77658-450 77641-722
3,5-Dimethoxy-N-methylaniline ≥97%
3,5-Dimethoxy-N-methylaniline

Precaución: For Research Use Only

Formula: 167.205 g/mol
MW: C₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD11152001
CAS Number: 118684-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR