Tert-butyl 4-formyl-3-methyl-benzoate ≥97%

Supplier: AOBChem USA

Warning

28372-5G 28372-25G 28372-1G 28372-500MG
77628-410EA 1484.23 USD
77628-410 77635-600 77637-758 77649-788
Tert-butyl 4-formyl-3-methyl-benzoate ≥97%
Tert-butyl 4-formyl-3-methyl-benzoate

Precaución: For Research Use Only

Formula: 220.264 g/mol
MW: C₁₃H₁₆O₃
Storage Temperature: Ambient
CAS Number: 1187028-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR