2-Bromo-4-(1-methyl-1-phenylethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

283382-5G 283382-25G 283382-1G 283382-500MG
77628-408EA 1484.23 USD
77628-408 77658-446 77645-708 77645-710
2-Bromo-4-(1-methyl-1-phenylethyl)phenol ≥97%
2-Bromo-4-(1-methyl-1-phenylethyl)phenol

Precaución: For Research Use Only

Formula: 291.18 g/mol
MW: C₁₅H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD34884246
CAS Number: 88882-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR