1,4-Diethyl-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

283081-5G 283081-1G 283081-25G 283081-500MG
77632-446EA 1207 USD
77632-446 77637-752 77658-438 77645-704
1,4-Diethyl-2-iodobenzene ≥95%
1,4-Diethyl-2-iodobenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₃I
Storage Temperature: Ambient
MDL Number: MFCD22491858
CAS Number: 1369920-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR