Cyclopropyl(2,5-diethylphenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

283044-5G 283044-500MG 283044-25G 283044-1G
77632-444EA 904 USD
77632-444 77652-588 77658-436 77641-712
Cyclopropyl(2,5-diethylphenyl)methanone ≥95%
Cyclopropyl(2,5-diethylphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₄H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD19669013
CAS Number: 1339873-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR