2-Chloro-1-(2,5-diethylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

283033-5G 283033-1G 283033-500MG 283033-25G
77634-612EA 575 USD
77634-612 77637-750 77652-586 77654-684
2-Chloro-1-(2,5-diethylphenyl)ethanone ≥95%
2-Chloro-1-(2,5-diethylphenyl)ethanone

Precaución: For Research Use Only

Formula: C₁₂H₁₅ClO
Storage Temperature: Ambient
CAS Number: 164165-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR