2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine, 4-Methyl-2-aminopyrimidine-5-boronic acid, pinacol ester

28288-5G 28288-1G 28288-25G
77634-610EA 225 USD
77634-610 77637-748 77654-682
2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester ≥97%
2-Amino-4-methyl-5-pyrimidineboronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₁H₁₈BN₃O₂
MW: 235.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18072554
CAS Number: 944401-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR