4-Methoxy-3-tricyclo[3.3.1.13,7]Dec-1-ylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

282875-5G 282875-25G 282875-1G 282875-500MG
77628-404EA 1484.23 USD
77628-404 77635-598 77637-746 77645-702
4-Methoxy-3-tricyclo[3.3.1.13,7]Dec-1-ylbenzaldehyde ≥95%
4-Methoxy-3-tricyclo[3.3.1.13,7]Dec-1-ylbenzaldehyde

Precaución: For Research Use Only

Formula: 270.37 g/mol
MW: C₁₈H₂₂O₂
Storage Temperature: Ambient
CAS Number: 107430-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR